Search
Search

ABI3| Gene Symbol: ABI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583598.2 | XP_005583655.1 | 3 - 4 | 952 | 88 | |
| XM_005583599.2 | XP_005583656.1 | 3 - 4 | 602 | 88 | ||
| XM_005583600.2 | XP_005583657.1 | 3 - 4 | 934 | 88 | ||
Set Membership: |
|