Search
Search

DGKH| Gene Symbol: DGKH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585737.2 | XP_005585794.1 | 12 - 13 | 1480 | 90 | |
| XM_005585738.2 | XP_005585795.1 | 12 - 13 | 1480 | 90 | ||
| XM_005585740.2 | XP_005585797.1 | 9 - 10 | 3553 | 90 | ||
| XM_015439201.1 | XP_015294687.1 | 9 - 10 | 1146 | 90 | ||
| XM_015439202.1 | XP_015294688.1 | 12 - 13 | 1480 | 90 | ||
| XM_015439203.1 | XP_015294689.1 | 5 - 6 | 629 | 90 | ||
Set Membership: |
|