Search
Search

NUP58| Gene Symbol: NUP58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585509.2 | XP_005585566.1 | 7 - 8 | 1378 | 114 | |
| XM_005585510.2 | XP_005585567.1 | 6 - 7 | 924 | 114 | ||
| XM_005585511.2 | XP_005585568.1 | 7 - 8 | 917 | 114 | ||
| XM_015439076.1 | XP_015294562.1 | 7 - 8 | 1378 | 114 | ||
| GenBank mRNA | AB170405.1 | - | 7 - 8 | 866 | 114 | |
Set Membership: |
|