Search
Search

CLPB| Gene Symbol: CLPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283620.1 | NP_001270549.1 | 12 - 13 | 1655 | 108 | |
| XM_005578997.2 | XP_005579054.1 | 13 - 14 | 1779 | 108 | ||
| XM_005578998.2 | XP_005579055.1 | 12 - 13 | 1688 | 108 | ||
| XM_005579000.2 | XP_005579057.1 | 11 - 12 | 1602 | 108 | ||
| GenBank mRNA | AB169030.1 | - | 12 - 13 | 1655 | 108 | |
| AB169361.1 | - | 13 - 14 | 1651 | 108 | ||
Set Membership: |
|