Search
Search

IKBKB| Gene Symbol: IKBKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563202.2 | XP_005563259.1 | 13 - 14 | 1549 | 105 | |
| XM_005563204.2 | XP_005563261.1 | 12 - 13 | 1454 | 105 | ||
| XM_015454727.1 | XP_015310213.1 | 13 - 14 | 1549 | 105 | ||
| XM_015454728.1 | XP_015310214.1 | 13 - 14 | 1512 | 105 | ||
| XM_015454729.1 | XP_015310215.1 | 12 - 13 | 1399 | 105 | ||
| XM_015454730.1 | XP_015310216.1 | 12 - 13 | 1454 | 105 | ||
Set Membership: |
|