Search
Search

CSMD1| Gene Symbol: CSMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562521.2 | XP_005562578.1 | 3 - 4 | 1044 | 80 | |
| XM_015454741.1 | XP_015310227.1 | 3 - 4 | 1044 | 80 | ||
Set Membership: |
|