Search
Search

TMEM8B| Gene Symbol: TMEM8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581342.2 | XP_005581399.1 | 9 - 10 | 1989 | 87 | |
| XM_005581343.2 | XP_005581400.1 | 9 - 10 | 1991 | 87 | ||
| XM_005581344.2 | XP_005581401.1 | 8 - 9 | 1183 | 87 | ||
| XM_015437009.1 | XP_015292495.1 | 9 - 10 | 2030 | 87 | ||
| XM_015437010.1 | XP_015292496.1 | 10 - 11 | 1732 | 87 | ||
| XM_015437011.1 | XP_015292497.1 | 8 - 9 | 1187 | 87 | ||
| XM_015437012.1 | XP_015292498.1 | 10 - 11 | 1732 | 87 | ||
Set Membership: |
|