Search
Search

PEX16| Gene Symbol: PEX16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578064.2 | XP_005578121.1 | 9 - 10 | 1975 | 59 | |
| XM_005578065.2 | XP_005578122.1 | 9 - 10 | 1975 | 59 | ||
| XM_005578066.2 | XP_005578123.1 | 9 - 10 | 1924 | 59 | ||
| XM_005578067.2 | XP_005578124.1 | 9 - 10 | 1924 | 59 | ||
| XM_005578068.2 | XP_005578125.1 | 9 - 10 | 1924 | 59 | ||
| XM_015434861.1 | XP_015290347.1 | 9 - 10 | 1975 | 59 | ||
Set Membership: |
|