Search
Search

KCNJ16| Gene Symbol: KCNJ16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584806.2 | XP_005584863.1 | 2 | 2614 | 120 | |
| XM_005584807.2 | XP_005584864.1 | 2 | 3051 | 120 | ||
| XM_005584808.2 | XP_005584865.1 | 2 | 2557 | 120 | ||
| XM_015438774.1 | XP_015294260.1 | 2 | 3260 | 120 | ||
| XM_015438775.1 | XP_015294261.1 | 2 | 4149 | 120 | ||
| XM_015438776.1 | XP_015294262.1 | 2 | 2631 | 120 | ||
Set Membership: |
|