Search
Search

WWOX| Gene Symbol: WWOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592616.2 | XP_005592673.1 | 1 - 2 | 746 | 94 | |
| XM_005592619.2 | XP_005592676.1 | 1 - 2 | 746 | 94 | ||
| XM_015443210.1 | XP_015298696.1 | 1 - 2 | 746 | 94 | ||
Set Membership: |
|