Search
Search

LYZL4| Gene Symbol: LYZL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546757.2 | XP_005546814.1 | 4 - 5 | 625 | 114 | |
| XM_005546758.2 | XP_005546815.1 | 4 - 5 | 530 | 114 | ||
Set Membership: |
|