Search
Search

SLC23A2| Gene Symbol: SLC23A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568408.2 | XP_005568465.1 | 14 - 15 | 2146 | 62 | |
| XM_005568409.2 | XP_005568466.1 | 13 - 14 | 3567 | 62 | ||
| XM_015457823.1 | XP_015313309.1 | 14 - 15 | 1781 | 62 | ||
| XM_015457824.1 | XP_015313310.1 | 13 - 14 | 1655 | 62 | ||
Set Membership: |
|