Search
Search

SLC35C2| Gene Symbol: SLC35C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569226.2 | XP_005569283.1 | 3 - 4 | 610 | 65 | |
| XM_005569227.2 | XP_005569284.1 | 3 - 4 | 613 | 65 | ||
| XM_005569229.2 | XP_005569286.1 | 3 - 4 | 542 | 65 | ||
| XM_005569230.2 | XP_005569287.1 | 3 - 4 | 542 | 65 | ||
| XM_015429902.1 | XP_015285388.1 | 3 - 4 | 613 | 65 | ||
| XM_015429903.1 | XP_015285389.1 | 3 - 4 | 613 | 65 | ||
Set Membership: |
|