Search
Search

INO80D| Gene Symbol: INO80D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574035.2 | XP_005574092.1 | 3 - 4 | 620 | 124 | |
| XM_005574036.2 | XP_005574093.1 | 3 - 4 | 551 | 124 | ||
| XM_005574038.2 | XP_005574095.1 | 3 - 4 | 540 | 124 | ||
| XM_015432621.1 | XP_015288107.1 | 3 - 4 | 401 | 124 | ||
| XM_015432622.1 | XP_015288108.1 | 3 - 4 | 450 | 124 | ||
| XM_015432623.1 | XP_015288109.1 | 3 - 4 | 389 | 124 | ||
Set Membership: |
|