Search
Search

SPATA20| Gene Symbol: SPATA20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583688.2 | XP_005583745.1 | 6 - 7 | 758 | 80 | |
| XM_005583689.1 | XP_005583746.1 | 5 - 6 | 727 | 80 | ||
Set Membership: |
|