Search
Search

LRRC27| Gene Symbol: LRRC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566766.2 | XP_005566823.1 | 8 - 9 | 1479 | 86 | |
| XM_005566767.2 | XP_005566824.1 | 8 - 9 | 1605 | 86 | ||
| XM_005566768.2 | XP_005566825.1 | 7 - 8 | 1486 | 86 | ||
| XM_005566769.2 | XP_005566826.1 | 6 - 7 | 862 | 86 | ||
Set Membership: |
|