Search
Search

RNF144B| Gene Symbol: RNF144B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553940.2 | XP_005553997.1 | 7 - 8 | 1108 | 83 | |
| XM_005553941.2 | XP_005553998.1 | 7 - 8 | 1001 | 83 | ||
Set Membership: |
|