Search
Search

UBE2U| Gene Symbol: UBE2U | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285106.1 | NP_001272035.1 | 6 - 7 | 930 | 80 | |
| XM_015452091.1 | XP_015307577.1 | 6 - 7 | 733 | 80 | ||
| GenBank mRNA | AB072767.1 | - | 6 - 7 | 930 | 80 | |
Set Membership: |
|