Search
Search

DOCK2| Gene Symbol: DOCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015452237.1 | XP_015307723.1 | 46 - 47 | 4813 | 77 | |
| XM_015452238.1 | XP_015307724.1 | 45 - 46 | 4717 | 77 | ||
Set Membership: |
|