Search
Search

CAPN14| Gene Symbol: CAPN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576185.2 | XP_005576242.1 | 17 - 18 | 2047 | 73 | |
| XM_005576187.2 | XP_005576244.1 | 17 - 18 | 2047 | 73 | ||
| XM_005576188.2 | XP_005576245.1 | 18 - 19 | 1912 | 73 | ||
| XM_015433829.1 | XP_015289315.1 | 17 - 18 | 4004 | 73 | ||
Set Membership: |
|