| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900004865.mm.1 |
NM_027397.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004865.mm.1 |
NM_027397.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004865.mm.1 |
TC0900000675.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004866.mm.1 |
NM_027397.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004866.mm.1 |
NM_027397.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004866.mm.1 |
TC0900000675.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004867.mm.1 |
NM_027397.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004867.mm.1 |
NM_027397.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004867.mm.1 |
TC0900000675.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004868.mm.1 |
NM_027397.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004868.mm.1 |
NM_027397.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004868.mm.1 |
TC0900000675.mm.1 |
MTA TC |