| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22603837 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22603838 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22603834 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22603835 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22603836 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2111237 |
BC019792.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2111237 |
AK020419.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2111237 |
AK159085.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2111237 |
AK205282.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2111237 |
AK213933.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569648 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
63339 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
151479 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
193732 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
669413 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
817606 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
573446 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
966621 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
138982 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
841967 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
927989 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
441726 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
45781 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428915 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
842799 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
35109 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310493 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914378 |
AK007453.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914378 |
AK035739.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914378 |
BC004569.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914378 |
BC019996.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914378 |
NM_028048.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
195000 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
130250 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
1221 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
80905 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P142630 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135649 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2111237 |
BC019792.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2111237 |
AK020419.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2111237 |
AK159085.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2111237 |
AK205282.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2111237 |
AK213933.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603866 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603846 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603860 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603857 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603865 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603856 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603839 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603842 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603835 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603847 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603853 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603870 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603868 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603851 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603864 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603862 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603841 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603849 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603858 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603852 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603844 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603838 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603836 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603834 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22603871 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135649 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05104687 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00779532 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05104671 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P142630 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P111143 |
AK020419.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P111143 |
AK159353.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P111143 |
AK205282.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P111143 |
AK213933.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P111143 |
AK218207.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104841 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P142630 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135649 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P00779532 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02535795 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02535776 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02535765 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22603837 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22603836 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22603835 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22603834 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22603838 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603847 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603985 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604033 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603983 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604084 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604088 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603968 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603884 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604009 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604078 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604026 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603920 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603903 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604017 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603990 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603861 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604069 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603926 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604071 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603832 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603892 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603965 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603839 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604058 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604065 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603842 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603916 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604045 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603980 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603876 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603947 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604010 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603911 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604081 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603976 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604008 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604057 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603937 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603932 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603943 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603857 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603898 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604031 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603979 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604064 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603894 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603836 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603851 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604002 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603981 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604070 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603945 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603844 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603846 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603938 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604006 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603915 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603879 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604025 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604015 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603914 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603866 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604022 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603970 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603956 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603966 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604041 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603858 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603870 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603972 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603860 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603978 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604000 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604013 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603835 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604049 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603838 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603862 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603975 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603874 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603912 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603896 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604063 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603853 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603962 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603929 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603887 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603952 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603834 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603954 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603881 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604054 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603841 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603864 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604042 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604007 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603924 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603994 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604029 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604086 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603868 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604067 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604074 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604039 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604043 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603923 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603889 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603982 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603852 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603993 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22604046 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603907 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603949 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603902 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603849 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603909 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22603930 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603943 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02396931 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603847 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603844 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604065 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603872 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02536285 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02536296 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603841 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603937 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603864 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603849 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603938 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603941 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02347780 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604033 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603929 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604026 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603933 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604074 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603881 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02480101 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603861 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604029 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603879 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603935 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02536294 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603860 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603932 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603871 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02372427 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604070 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603846 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603858 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604019 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603839 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603884 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02372422 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603945 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604039 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603834 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603842 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604067 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02535780 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603876 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603866 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02372407 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603873 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604025 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604064 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02396927 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604069 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02536288 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603851 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02535807 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02535784 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603835 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22603947 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22604071 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02372421 |
- |
- |