Search
Search

Magoh| Gene Symbol: Magoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282737.1 | NP_001269666.1 | 1 - 2 | 193 | 68 | |
| GenBank mRNA | AF007862.1 | - | 1 - 2 | 180 | 68 | |
| AF035939.1 | - | 1 - 2 | 140 | 68 | ||
| AK011512.1 | - | 1 - 2 | 132 | 68 | ||
| BC018176.1 | - | 1 - 2 | 131 | 3587774 | 68 | |
| BY134232.1 | - | 1 - 2 | 194 | 68 | ||
Set Membership: |
|