Search
Search

Rabggta| Gene Symbol: Rabggta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019519.2 | NP_062392.1 | 9 - 10 | 1340 | 88 | |
| GenBank mRNA | AF127656.1 | - | 10 - 11 | 1188 | 88 | |
| AF127657.1 | - | 9 - 10 | 1131 | 88 | ||
| AF127658.1 | - | 9 - 10 | 1340 | 88 | ||
| AF127659.1 | - | 9 - 10 | 1340 | 88 | ||
| AF127662.1 | - | 10 - 11 | 1259 | 88 | ||
| AK002625.1 | - | 10 - 11 | 1122 | 88 | ||
| AK146917.1 | - | 10 - 11 | 1211 | 88 | ||
Set Membership: |
|