Search
Search
| Gene Symbol: Dsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291804.1 | NP_001278733.1 | 1 - 2 | 283 | 84 | |
| NM_013504.5 | NP_038532.3 | 1 - 2 | 283 | 84 | ||
| GenBank mRNA | AK036893.1 | - | 1 - 2 | 169 | 84 | |
| AK079441.1 | - | 1 - 2 | 284 | 84 | ||
| AK081826.1 | - | 1 - 2 | 191 | 84 | ||
| BC109161.1 | - | 1 - 2 | 120 | 40047883 | 84 | |
| BC109162.1 | - | 1 - 2 | 120 | 84 | ||
| X97986.1 | - | 1 - 2 | 217 | 84 | ||
Set Membership: |
|