Search
Search
| Gene Symbol: Ubl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122873.1 | NP_001116345.1 | 3 - 4 | 409 | 60 | |
| NM_027086.3 | NP_081362.2 | 3 - 4 | 357 | 60 | ||
| GenBank mRNA | AK009025.1 | - | 3 - 4 | 358 | 60 | |
| AK154496.1 | - | 3 - 4 | 367 | 60 | ||
| BC016456.1 | - | 2 - 3 | 343 | 4912715 | 60 | |
| BC094047.1 | - | 2 - 3 | 346 | 5253101 | 60 | |
| CX203425.1 | - | 3 - 4 | 387 | 60 | ||
Set Membership: |
|