Search
Search

Meaf6| Gene Symbol: Meaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290701.1 | NP_001277630.1 | 3 - 4 | 344 | 76 | |
| NM_027310.4 | NP_081586.1 | 3 - 4 | 344 | 76 | ||
| GenBank mRNA | AK009177.1 | - | 3 - 4 | 332 | 76 | |
| AK012867.1 | - | 3 - 4 | 316 | 76 | ||
| BC061024.1 | - | 3 - 4 | 314 | 76 | ||
| BC108404.1 | - | 3 - 4 | 315 | 30135798 | 76 | |
Set Membership: |
|