Search
Search

Crocc| Gene Symbol: Crocc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145958.1 | NP_001139430.1 | 29 - 30 | 4673 | 100 | |
| NM_172122.2 | NP_742120.2 | 30 - 31 | 4926 | 100 | ||
| GenBank mRNA | AF527975.1 | - | 30 - 31 | 4926 | 100 | |
| AK032335.1 | - | 2 - 3 | 250 | 100 | ||
| AK122285.1 | - | 21 - 22 | 4169 | 100 | ||
| BC027090.1 | - | 3 - 4 | 467 | 100 | ||
| BC034536.1 | - | 7 - 8 | 1831 | 100 | ||
| BC036729.1 | - | 15 - 16 | 2568 | 100 | ||
| BC054054.1 | - | 28 - 29 | 4654 | 6394697 | 100 | |
Set Membership: |
|