Search
Search
| Gene Symbol: Dnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038619.1 | NP_001033708.1 | 6 - 7 | 1056 | 65 | |
| NM_172646.2 | NP_766234.1 | 6 - 7 | 1060 | 65 | ||
| GenBank mRNA | AK036199.1 | - | 6 - 7 | 1021 | 65 | |
| AK049724.1 | - | 6 - 7 | 1061 | 65 | ||
| AK147678.1 | - | 6 - 7 | 1000 | 65 | ||
| BC141143.1 | - | 6 - 7 | 904 | 9055755 | 65 | |
| BC141144.1 | - | 6 - 7 | 904 | 9055756 | 65 | |
Set Membership: |
|