Search
Search

Ubash3b| Gene Symbol: Ubash3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176860.5 | NP_789830.1 | 7 - 8 | 1146 | 62 | |
| GenBank mRNA | AB075602.1 | - | 7 - 8 | 1141 | 62 | |
| AK034450.1 | - | 7 - 8 | 1337 | 62 | ||
| AK035764.1 | - | 7 - 8 | 1147 | 62 | ||
| AK133948.1 | - | 7 - 8 | 1144 | 62 | ||
| AK150909.1 | - | 7 - 8 | 1713 | 62 | ||
| AK151721.1 | - | 7 - 8 | 1714 | 62 | ||
| AK151895.1 | - | 7 - 8 | 1347 | 62 | ||
| AK152013.1 | - | 7 - 8 | 1347 | 62 | ||
| AK154576.1 | - | 7 - 8 | 1368 | 62 | ||
| BC053436.1 | - | 7 - 8 | 1118 | 6510702 | 62 | |
Set Membership: |
|