Search
Search

Ric8b| Gene Symbol: Ric8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013441.2 | NP_001013459.1 | 4 - 5 | 959 | 82 | |
| NM_183172.2 | NP_898995.1 | 4 - 5 | 959 | 82 | ||
| GenBank mRNA | AK132369.1 | - | 4 - 5 | 863 | 82 | |
| AK137531.1 | - | 3 - 4 | 757 | 82 | ||
| AY940666.1 | - | 4 - 5 | 928 | 82 | ||
| AY940667.1 | - | 4 - 5 | 916 | 82 | ||
| BC051080.1 | - | 4 - 5 | 956 | 6490136 | 82 | |
Set Membership: |
|