Search
Search

Phf14| Gene Symbol: Phf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168382.1 | NP_001161854.1 | 4 - 5 | 1442 | 157 | |
| NM_029404.3 | NP_083680.2 | 4 - 5 | 1439 | 157 | ||
| GenBank mRNA | AK016517.1 | - | 4 - 5 | 1442 | 157 | |
| BC040236.1 | - | 4 - 5 | 1405 | 3585881 | 157 | |
Set Membership: |
|