Search
Search
| Gene Symbol: Msh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF107352.1 | - | 6 - 7 | 569 | 68 | |
| AF146227.1 | - | 5 - 6 | 777 | 68 | ||
| AK017308.1 | - | 1 - 2 | 691 | 68 | ||
| AK042083.1 | - | 6 - 7 | 554 | 68 | ||
| BC030034.1 | - | 4 - 5 | 857 | 68 | ||
| BC141113.1 | - | 5 - 6 | 697 | 9055725 | 68 | |
| BC145406.1 | - | 5 - 6 | 697 | 68 | ||
| BC145407.1 | - | 5 - 6 | 697 | 68 | ||
Set Membership: |
|