Search
Search

Atcay| Gene Symbol: Atcay | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178662.3 | NP_848777.1 | 2 - 3 | 418 | 56 | |
| GenBank mRNA | AK028298.1 | - | 2 - 3 | 425 | 56 | |
| AK038483.1 | - | 2 - 3 | 428 | 56 | ||
| AK038990.1 | - | 2 - 3 | 419 | 56 | ||
| AK039411.1 | - | 2 - 3 | 426 | 56 | ||
| AK086723.1 | - | 2 - 3 | 460 | 56 | ||
| AK162959.1 | - | 2 - 3 | 426 | 56 | ||
| AY349150.1 | - | 2 - 3 | 383 | 56 | ||
| BC048903.1 | - | 2 - 3 | 402 | 6491141 | 56 | |
Set Membership: |
|