Search
Search

Clstn3| Gene Symbol: Clstn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153508.4 | NP_705728.1 | 14 - 15 | 2545 | 72 | |
| GenBank mRNA | AJ278486.1 | - | 14 - 15 | 2412 | 72 | |
| AK032336.1 | - | 14 - 15 | 2487 | 72 | ||
| AK194842.1 | - | 2 - 3 | 289 | 72 | ||
| BC041781.1 | - | 14 - 15 | 2244 | 72 | ||
| BC055054.1 | - | 14 - 15 | 2477 | 6493096 | 72 | |
Set Membership: |
|