Search
Search

Foxj3| Gene Symbol: Foxj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290696.1 | NP_001277625.1 | 8 - 9 | 1454 | 63 | |
| NM_172699.3 | NP_766287.1 | 9 - 10 | 1556 | 63 | ||
| GenBank mRNA | AK082840.1 | - | 9 - 10 | 1557 | 63 | |
| AK129271.1 | - | 8 - 9 | 1427 | 63 | ||
| AK139508.1 | - | 9 - 10 | 1490 | 63 | ||
| BC058231.1 | - | 9 - 10 | 1529 | 6314981 | 63 | |
Set Membership: |
|