Search
Search
| Gene Symbol: Hgh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021555.2 | NP_067530.2 | 4 - 5 | 966 | 84 | |
| GenBank mRNA | AB041552.1 | - | 4 - 5 | 966 | 84 | |
| AK085815.1 | - | 4 - 5 | 960 | 84 | ||
| AK178524.1 | - | 2 - 3 | 245 | 84 | ||
| BC061251.1 | - | 4 - 5 | 930 | 30249656 | 84 | |
Set Membership: |
|