Search
Search

Tbc1d1| Gene Symbol: Tbc1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289514.1 | NP_001276443.1 | 11 - 12 | 1793 | 82 | |
| NM_019636.3 | NP_062610.2 | 12 - 13 | 2374 | 82 | ||
| GenBank mRNA | AK122445.1 | - | 14 - 15 | 3462 | 82 | |
| AK135665.1 | - | 11 - 12 | 1965 | 82 | ||
| AK135892.1 | - | 11 - 12 | 2058 | 82 | ||
| AK148356.1 | - | 11 - 12 | 1750 | 82 | ||
| AK154591.1 | - | 13 - 14 | 2410 | 82 | ||
| AK159256.1 | - | 12 - 13 | 2370 | 82 | ||
| AK164923.1 | - | 8 - 9 | 1078 | 82 | ||
| BC004675.1 | - | 1 - 2 | 32 | 82 | ||
| BC117931.1 | - | 11 - 12 | 2042 | 40097712 | 82 | |
| U33005.1 | - | 12 - 13 | 2330 | 82 | ||
Set Membership: |
|