Search
Search
| Gene Symbol: Arnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037737.2 | NP_001032826.1 | 13 - 14 | 1390 | 114 | |
| NM_009709.4 | NP_033839.2 | 12 - 13 | 1345 | 114 | ||
| GenBank mRNA | AK028546.1 | - | 12 - 13 | 1332 | 114 | |
| AK040475.1 | - | 13 - 14 | 1392 | 114 | ||
| AK141362.1 | - | 13 - 14 | 1261 | 114 | ||
| AK145415.1 | - | 12 - 13 | 1300 | 114 | ||
| AK152473.1 | - | 13 - 14 | 1372 | 114 | ||
| AK153355.1 | - | 13 - 14 | 1372 | 114 | ||
| AK220458.1 | - | 9 - 10 | 1521 | 114 | ||
| BC012870.1 | - | 13 - 14 | 1352 | 3964932 | 114 | |
| U10325.1 | - | 13 - 14 | 1254 | 114 | ||
Set Membership: |
|