Search
Search

Epha4| Gene Symbol: Epha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007936.3 | NP_031962.2 | 7 - 8 | 1665 | 75 | |
| GenBank mRNA | AK028509.1 | - | 7 - 8 | 1658 | 75 | |
| AK132203.1 | - | 7 - 8 | 1664 | 75 | ||
| AK147698.1 | - | 7 - 8 | 1661 | 75 | ||
| BC004782.1 | - | 7 - 8 | 1580 | 3588285 | 75 | |
| BC052164.1 | - | 7 - 8 | 1637 | 6512978 | 75 | |
| S57168.1 | - | 7 - 8 | 1665 | 75 | ||
| X65138.1 | - | 7 - 8 | 1665 | 75 | ||
Set Membership: |
|