Search
Search

Fbxo38| Gene Symbol: Fbxo38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134136.3 | NP_598897.2 | 9 - 10 | 1264 | 117 | |
| GenBank mRNA | AK031096.1 | - | 9 - 10 | 1214 | 117 | |
| AK031347.1 | - | 9 - 10 | 1195 | 117 | ||
| AK034761.1 | - | 4 - 5 | 448 | 117 | ||
| AK089046.1 | - | 9 - 10 | 1236 | 117 | ||
| AY267463.1 | - | 9 - 10 | 1197 | 117 | ||
| BC056348.1 | - | 9 - 10 | 1216 | 5721407 | 117 | |
Set Membership: |
|