Search
Search

Zfp536| Gene Symbol: Zfp536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039356.1 | - | 1 - 2 | 129 | 73 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | teashirt zinc finger family member 3 | Tshz3 |
A630038G13Rik, Tsh3, Zfp537, mKIAA1474, teashirt3 |
-943.0kb | Anti-sense | Chr. 7 - 36697742 - 36773553 on NCBI Build GRCm38 |
| Downstream |
URI1, prefoldin-like chaperone |
Uri1 |
C80913, NNX3, Rmp |
186.5kb | Sense | Chr. 7 - 37959992 - 38023575 on NCBI Build GRCm38 |
Set Membership: |
|