Search
Search

Pcsk2| Gene Symbol: Pcsk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008792.4 | NP_032818.1 | 9 - 10 | 1451 | 70 | |
| GenBank mRNA | AF483512.1 | - | 9 - 10 | 1093 | 70 | |
| AF483513.1 | - | 9 - 10 | 1093 | 70 | ||
| AK133620.1 | - | 9 - 10 | 1452 | 70 | ||
| AK158493.1 | - | 9 - 10 | 1399 | 70 | ||
| AK161585.1 | - | 9 - 10 | 1392 | 70 | ||
| BC052013.1 | - | 9 - 10 | 1368 | 5707923 | 70 | |
| BC057348.1 | - | 9 - 10 | 1406 | 6827695 | 70 | |
| M55669.1 | - | 9 - 10 | 1177 | 70 | ||
Set Membership: |
|