| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200007140.mm.1 |
NM_026361.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007140.mm.1 |
NM_026361.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007140.mm.1 |
NM_175464.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007140.mm.1 |
NM_175464.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007140.mm.1 |
TC0200000888.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200007143.mm.1 |
NM_026361.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007143.mm.1 |
NM_026361.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007143.mm.1 |
NM_175464.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007143.mm.1 |
NM_175464.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007143.mm.1 |
TC0200000888.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200007141.mm.1 |
NM_026361.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007141.mm.1 |
NM_026361.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007141.mm.1 |
NM_175464.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007141.mm.1 |
NM_175464.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007141.mm.1 |
TC0200000888.mm.1 |
MTA TC |