Search
Search

Dennd5a| Gene Symbol: Dennd5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021494.1 | NP_067469.1 | 12 - 13 | 2674 | 97 | |
| GenBank mRNA | AJ245569.1 | - | 11 - 12 | 2580 | 97 | |
| AK048609.1 | - | 3 - 4 | 328 | 97 | ||
| AK084641.1 | - | 6 - 7 | 2153 | 97 | ||
| BC022119.1 | - | 8 - 9 | 1327 | 5067385 | 97 | |
| BC060230.1 | - | 12 - 13 | 2674 | 6840562 | 97 | |
| L40894.1 | - | 3 - 4 | 322 | 97 | ||
Set Membership: |
|