Search
Search
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173767.3 | NP_776128.2 | 8 - 9 | 1275 | 62 | |
| GenBank mRNA | AK014452.1 | - | 7 - 8 | 1036 | 62 | |
| AK050143.1 | - | 5 - 6 | 770 | 62 | ||
| AK204816.1 | - | 2 - 3 | 229 | 62 | ||
| AK211253.1 | - | 2 - 3 | 223 | 62 | ||
| BC026405.1 | - | 8 - 9 | 1162 | 4211657 | 62 | |
| BC089586.1 | - | 5 - 6 | 779 | 62 | ||
| BC139196.1 | - | 5 - 6 | 663 | 8862218 | 62 | |
| BC139197.1 | - | 5 - 6 | 663 | 8862219 | 62 | |
| DQ205645.1 | - | 7 - 8 | 988 | 62 | ||
Set Membership: |
|