Search
Search

Palb2| Gene Symbol: Palb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081238.2 | NP_001074707.1 | 2 - 3 | 291 | 123 | |
| NM_001289842.1 | NP_001276771.1 | 2 - 3 | 291 | 123 | ||
| NM_001289844.1 | NP_001276773.1 | 2 - 3 | 291 | 123 | ||
| GenBank mRNA | AK028653.1 | - | 2 - 3 | 265 | 123 | |
| AK154227.1 | - | 2 - 3 | 263 | 123 | ||
| AK156701.1 | - | 2 - 3 | 210 | 123 | ||
| AK157382.1 | - | 2 - 3 | 210 | 123 | ||
| BC066140.1 | - | 2 - 3 | 189 | 30459312 | 123 | |
Set Membership: |
|