Search
Search

Phf10| Gene Symbol: Phf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024250.4 | NP_077212.3 | 7 - 8 | 873 | 67 | |
| GenBank mRNA | AB221576.1 | - | 5 - 6 | 540 | 67 | |
| AK007873.1 | - | 7 - 8 | 853 | 67 | ||
| BC002206.1 | - | 5 - 6 | 607 | 3489870 | 67 | |
| CA322732.1 | - | 4 - 5 | 431 | 67 | ||
| CD363469.1 | - | 1 - 2 | 130 | 67 | ||
Set Membership: |
|