Search
Search
| Gene Symbol: Ric3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038624.1 | NP_001033713.1 | 5 - 6 | 706 | 83 | |
| GenBank mRNA | AK134663.1 | - | 5 - 6 | 688 | 83 | |
| AK138641.1 | - | 5 - 6 | 702 | 83 | ||
| BC059258.1 | - | 4 - 5 | 448 | 6408852 | 83 | |
Set Membership: |
|