Search
Search

Zdhhc9| Gene Symbol: Zdhhc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172465.4 | NP_766053.1 | 4 - 5 | 834 | 86 | |
| GenBank mRNA | AK032233.1 | - | 4 - 5 | 816 | 86 | |
| AK079248.1 | - | 4 - 5 | 802 | 86 | ||
| BC042618.1 | - | 4 - 5 | 693 | 4950217 | 86 | |
| BC090832.1 | - | 4 - 5 | 652 | 6401763 | 86 | |
Set Membership: |
|